Learn More
Macherey-Nagel™ Polygram CEL 300-25 UV254 Glass Plates
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Pack of 25
Specifications
Specifications
Quantity | 25 Pk. |
Type | Glass Plate |
Fluorescent Indicator | UV254 |
Thickness (Metric) | 0.25 mm |
Dimensions (L x W) | 20 x 20 cm |
Product Content Correction
Product Title
Macherey-Nagel™ Polygram CEL 300-25 UV254 Glass Plates >
Spot an opportunity for improvement?Share a Content Correction