Learn More
Vitlab™ Polyoxymethylene Forceps
Manufactured with POM for excellent dimensional stability. VITLab™ Polyoxymethylene Forcep have grooves on the outside for optimum handling and grip.
Brand: Vitlab™ 68199
This item is not returnable.
View return policy
Description
- For bottling and decanting
- Yello
- Blunt
- Elastic
- Very good resilience
Specifications
Round | |
145 mm | |
5/Pack | |
Forceps |
POM | |
Forceps | |
For autoclaving, bottling and decanting |
Product Content Correction
Product Title
Vitlab™ Polyoxymethylene Forceps > Tip Style: Rounded end; Length: 145mm
Spot an opportunity for improvement?Share a Content Correction