Learn More
Hirschmann™ Polypropylene 2-10 mL EM Dispenser
A bottle attachment dispenser for non-problematic watery and organic media which do not attack polypropylene.
Brand:  Hirschmann™ 9340200
Product Code. 10452491
  • 2515.00 DKK / Each
Explore more special offers
Description
Description
  • With variable volume configuration in appropriate steps
  • Certified conformity
  • With precision ground glass piston
  • Suitable for steam sterilization up to 121°C
  • A 32 screw thread
  • Delivery complete with 3 threaded adapters (A 28, S 40, A 45), 1 suction hose with union nut, 1 ejection unit and 1 fitting wrench
  • With individual serial number and calibration certificate
Specifications
Specifications
Dispenser
Dispensing liquids
2 to 10 mL