Learn More
Eppendorf™ Polypropylene Adaptor
Brand: Eppendorf™ 4960800040
70.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Adapter | |
Polypropylene | |
Bottle thread 25 mm | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Product Title
Eppendorf™ Polypropylene Adaptor > For Bottle thread 25 mm
Spot an opportunity for improvement?Share a Content Correction