Learn More
Eppendorf™ Polypropylene Adaptor
Brand: Eppendorf™ 4960800147
97.75 DKK valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Adapter | |
Polypropylene | |
Bottle thread GL 45 to S 40 | |
1 Ea. |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92% -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Product Content Correction
Product Title
Eppendorf™ Polypropylene Adaptor > For Bottle thread GL 45 to S 40
Spot an opportunity for improvement?Share a Content Correction