Learn More
Buerkle™ Polypropylene Barrel Screw Joint
Buerkle™ Polypropylene Barrel Screw Joint for secure connection in the barrel in three different thread models.
328.00 DKK - 447.00 DKK
Specifications
Material | Polypropylene |
---|---|
Type | Barrel Screw Joint |
For Use With | PTFE Barrel Pump |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10309780
|
Buerkle™
5600-3170 |
Mauser 2 in., Coarse Outer Thread |
408.00 DKK
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
10625571
|
Buerkle™
5600-3130 |
R 2 in., Fine Steel Outer Thread |
328.00 DKK
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
10126370
|
Buerkle™
5600-3180 |
Tri-Sure, Coarse Outer Thread |
447.00 DKK
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
Description
- Barrel screw joint made of PP for secure connection in the barrel in three different thread models
- For drawing off liquids that are not flammable
- The container connectors make decanting even easier and safer with their more secure connection to barrels
- The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange
Specifications
Polypropylene | |
Barrel Screw Joint | |
PTFE Barrel Pump |