Learn More
BRAND™ Polypropylene Bucket
Reinforced rim and handle offer sturdy control. Spout provides easy pouring and prevents spilling.
Brand:  BRAND™ 72378
279.41 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10111871
  • 337.00 DKK / Each
Only null left
In Stock
Description
  • Polypropylene construction
  • Reinforced rim and handle
  • Includes spout
  • Lid not included
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Translucent
1 Ea.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Polypropylene Bucket > Capacity: 15L