missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polypropylene: ISO Safety Sign - Direction arrow 45°, Top Right Corner, Green

Sign compliant to ISO 7010.

Brand:  Brady™ 814395

 View more versions of this product

Product Code. 15614267

  • 114.00 DKK / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polypropylene
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
Indoor and Outdoor
1.501 mm
-40°C to 120°C
Global
Polypropylene
315 mm
315 mm
Square
White on Green
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers