Learn More
Cole-Parmer™ Polypropylene Manifolds
Brand: Cole-Parmer™ 31522-19
This item is not returnable.
View return policy
Specifications
Manifold | |
Polypropylene | |
4.3 mm | |
25.4 mm |
1/4 in. | |
8 | |
155.9 mm | |
25.4 mm |
Product Content Correction
Product Title
Cole-Parmer™ Polypropylene Manifolds > Length: 155.9mm
Spot an opportunity for improvement?Share a Content Correction