Learn More
BRAND™ Polypropylene Photographic Trays
White stackable polypropylene (PP) photographic trays with ribbed bottom that improves stability.
Brand:  BRAND™ 156650
786.39 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10706711
  • 882.00 DKK / Each
In Stock
Explore more special offers
Description
  • White stackable polypropylene (PP) photographic trays
  • Ribbed bottom improves stability, reinforced rim, slightly sloping walls
  • Autoclavable (121°C)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
160 mm
Product Title
BRAND™ Polypropylene Photographic Trays > Height (Metric): 160 mm