missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ Polypropylene Reagent Bottles

Use these bottles for long-term storage of liquids. VITLAB™ Polypropylene Reagent Bottles feature a wider mouth for simple filling of powders and paste-like materials.

Brand:  Vitlab™ 101689

 View more versions of this product

Product Code. 12885733

  • 600.00 DKK / Pack of 20
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Transparent Reagent Bottles
  • Good chemical resistance
  • With polypropylene screw cap
  • Autoclavable at 121°C (2 bar) according to DIN EN 285
  • 5000mL size features one handle for easy transport and pouring
  • Suitable for foodstuffs
Specifications

Specifications

250 mL
Ideal for powders and paste-like materials
Polypropylene
Yes
Wide
Reagent Bottle
73 mm
Transparent
Screw cap
20 Pack
Autoclavable
132 mm
Polypropylene
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.