Learn More
Grant Instruments™ Polypropylene Spheres
Pack of 300 polypropylene spheres
Brand:  Grant Instruments™ PS20
872.93 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11466467
  • 897.00 DKK / Pack of 300
Only null left
Estimated Shipment: 26-08-2025
Description
Pack of 300 polypropylene spheres
Useful alternative to a lid, minimises evaporation and heat loss whilst allowing easy access to vessels in the bath; particularly useful for tall vessels
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bath Accessories
Polypropylene
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Grant Instruments™ Polypropylene Spheres > Weight: 0.75kg