Learn More
VITLAB™ Polypropylene Standard Joint Funnels
Fill multi-neck reaction flasks with standard neck joints. VITLAB™ Polypropylene Standard Joint Funnels are transparent, laterally flattened funnels that fit snugly into standard neck joints for fast, easy filling. Available for a variety of standard neck sizes.
Brand:  VITLAB™ 70594
 View more versions of this product
Product Code. 10509101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Suitable for the filling of liquid or powdered reagents into a reaction flask during a reaction
  • Laterally flattened for use with multi-neck flasks
  • Suitable for standard joint necks of various sizes
  • Transparent material for easy monitoring of filling progress
  • Constructed of durable polypropylene
    • Chemically resistant for use with liquid or powdered reagents
    • Temperature resistant and autoclavable
    • FDA-grade quality
Specifications
Specifications
50mm
95 mm
Thread size: NS 19/26
Multi-Neck Flasks and Single-Neck Flasks with Standard Joint Necks
10Pack
Polypropylene
Transparent
Suitable for the Filling of Liquid or Powdered Reagents Into a Reaction Flask During a Reaction
23mm
Standard Joint Funnel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VITLAB™ Polypropylene Standard Joint Funnels > Dimensions (L x W): 95 x 50mm