Learn More
VITLAB™ Polypropylene Standard Joint Funnels
Fill multi-neck reaction flasks with standard neck joints. VITLAB™ Polypropylene Standard Joint Funnels are transparent, laterally flattened funnels that fit snugly into standard neck joints for fast, easy filling. Available for a variety of standard neck sizes.
Brand:  VITLAB™ 70594
 View more versions of this product
Product Code. 10509101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Suitable for the filling of liquid or powdered reagents into a reaction flask during a reaction
  • Laterally flattened for use with multi-neck flasks
  • Suitable for standard joint necks of various sizes
  • Transparent material for easy monitoring of filling progress
  • Constructed of durable polypropylene
    • Chemically resistant for use with liquid or powdered reagents
    • Temperature resistant and autoclavable
    • FDA-grade quality
Specifications
Specifications
50mm
95 mm
Thread size: NS 19/26
Multi-Neck Flasks and Single-Neck Flasks with Standard Joint Necks
10Pack
Polypropylene
Transparent
Suitable for the Filling of Liquid or Powdered Reagents Into a Reaction Flask During a Reaction
23mm
Standard Joint Funnel
Certificates
Promotions
Promotions
Product Content Correction
Product Title
VITLAB™ Polypropylene Standard Joint Funnels > Dimensions (L x W): 95 x 50mm