missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Polypropylene Standard Joint Funnels

Fill multi-neck reaction flasks with standard neck joints. VITLAB™ Polypropylene Standard Joint Funnels are transparent, laterally flattened funnels that fit snugly into standard neck joints for fast, easy filling. Available for a variety of standard neck sizes.

Brand:  VITLAB™ 70494

 View more versions of this product

Product Code. 12826533

  • 311.00 DKK / Pack of 10
Estimated Shipment: 10-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Suitable for the filling of liquid or powdered reagents into a reaction flask during a reaction
  • Laterally flattened for use with multi-neck flasks
  • Suitable for standard joint necks of various sizes
  • Transparent material for easy monitoring of filling progress
  • Constructed of durable polypropylene
    • Chemically resistant for use with liquid or powdered reagents
    • Temperature resistant and autoclavable
    • FDA-grade quality
Specifications

Specifications

40mm
75 mm
Thread size: NS 14/23
Multi-Neck Flasks and Single-Neck Flasks with Standard Joint Necks
10Pack
Polypropylene
Transparent
Suitable for the Filling of Liquid or Powdered Reagents Into a Reaction Flask During a Reaction
17mm
Standard Joint Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VITLAB™ Polypropylene Standard Joint Funnels > Dimensions (L x W): 75 x 40mm; Quantity: 10 pack

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.