Learn More
Cole-Parmer™ Polypropylene Storage Container Lids
Use these lids for sample containers 06100-12 and 06100-14. Cole-Parmer™ Polypropylene Storage Container Lids and containers are chemical resistant to acids bases and most organic solvents.
Brand: Cole-Parmer™ 06100-16
This item is not returnable.
View return policy
Description
- 500 lids per case
- Made of polypropylene (PP)
- Resist most chemicals
- Containers sold separately
Specifications
500/Cs. | |
Storage Container Lid | |
Polypropylene | |
Sample Containers 06100-12 and 06100-14 |
Product Content Correction
Product Title
Cole-Parmer™ Polypropylene Storage Container Lids > For Use With: Sample Containers 06100-12 and 06100-14
Spot an opportunity for improvement?Share a Content Correction