Learn More
Gosselin™ Polypropylene Straight Specimen Containers
CORNING GOSSELIN™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
Brand:  Gosselin™ TP35C-030
 View more versions of this product
Product Code. 15408824
  • 775.36 DKK / Pack of 800
Description
  • Clear and break-resistant polypropylene (PP)
  • Available in different sizes and quantities
  • Color-coded screw caps
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Specimen Container
Polypropylene
Non-sterile
Clear
800/Cs.
No
Straight
60 mL
No Label
800/Pk.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Gosselin™ Polypropylene Straight Specimen Containers > 60mL