missing translation for 'onlineSavingsMsg'
Learn More
VITLAB™ Wide-Mouth Bottles with Screw Caps Product Code.: 10087602
Add to basket

VITLAB™ Wide-Mouth Bottles with Screw Caps

Product Code. 10087602
500 mL, Polypropylene, Autoclavable, 45
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Material:
LDPE
Polypropylene
Autoclavable:
Autoclavable
Not Autoclavable
Thread Size (Metric):
40
45
50
63
65
This item is not returnable. View return policy

Product Code. 10087602

Brand: VITLAB™ 93994

Add to basket
Add to basket
This item is not returnable. View return policy

Fill bottles quickly and easily. VITLAB™ Wide-Mouth Bottles with Screw Caps feature a wide-neck opening for simple filling. Bottles are constructed of transparent polypropylene or low-density polyethylene and feature strong screw-cap lids. Available in a variety of sizes.

Filling narrow-necked reagent bottles with liquids, powders and paste-like materials can be very difficult, even with a funnel. These economical and durable plastic reagent bottles feature a wide opening to make filling quick and easy.
  • Transparent media bottles ideal for storage of a variety of chemicals
  • Reagent bottles featuring screw caps with high-pitch GL threads for significant carrying power
  • Wider mouth bottles are ideal for easy bottle filling, especially of powders and paste-like materials
  • Choice of bottle/cap materials:
    • Polypropylene (PP):
      Good chemical and temperature resistance, autoclavable, ideal for long-term storage
    • Low-Density Polyethylene (LDPE):
      Flexible and resilient material with excellent resistance to many acids and alcohols

Specifications

Capacity (Metric) 500 mL
Color Transparent
For Use With (Application) Saving and Storage
Includes Screw cap
Material Polypropylene
No. per Pack 12 Bottles
Thread Size (Metric) 45
Autoclavable Autoclavable
Height (Metric) 166 mm
Product Type Wide Mouth Bottle
Diameter (Metric) 76 mm
Closure Type Screw Cap with GL Thread
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VITLAB™ Wide-Mouth Bottles with Screw Caps >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.