missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Wide-Mouth Bottles with Screw Caps
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Material:
LDPE
Polypropylene
Autoclavable:
Autoclavable
Not Autoclavable
Thread Size (Metric):
40
45
50
63
65
Description
Filling narrow-necked reagent bottles with liquids, powders and paste-like materials can be very difficult, even with a funnel. These economical and durable plastic reagent bottles feature a wide opening to make filling quick and easy.
- Transparent media bottles ideal for storage of a variety of chemicals
- Reagent bottles featuring screw caps with high-pitch GL threads for significant carrying power
- Wider mouth bottles are ideal for easy bottle filling, especially of powders and paste-like materials
- Choice of bottle/cap materials:
- Polypropylene (PP):
Good chemical and temperature resistance, autoclavable, ideal for long-term storage - Low-Density Polyethylene (LDPE):
Flexible and resilient material with excellent resistance to many acids and alcohols
- Polypropylene (PP):
Specifications
Specifications
Capacity (Metric) | 250 mL |
Color | Transparent |
For Use With (Application) | Saving and Storage |
Includes | Screw cap |
Material | Polypropylene |
No. per Pack | 12 Bottles |
Thread Size (Metric) | 45 |
Autoclavable | Autoclavable |
Height (Metric) | 146 mm |
Product Type | Wide Mouth Bottle |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VITLAB™ Wide-Mouth Bottles with Screw Caps >
Spot an opportunity for improvement?Share a Content Correction