missing translation for 'onlineSavingsMsg'
Learn More

Kappes Systeme™ Polystyrene Lid

Encompass
Encompass_Preferred

POA

Specifications

Type Bin Lid
Material Polystyrene
Color Transparent
For Use With RasterPlan Plastic Bin
For Use With (Application) Food Storage
View More Specs

Products 3
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
10218302
Encompass
View Documents
Kappes Systeme™
6060.00.0557
160 x 105 x 75 mm
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

10094052
Encompass_Preferred
View Documents
Kappes Systeme™
6060.00.0457
230 x 140 x 130 mm
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

10695633
Encompass
View Documents
Kappes Systeme™
6060.00.0357
290 x 140 x 130 mm
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

Specifications

Specifications

Bin Lid
Transparent
Food Storage
Polystyrene
RasterPlan Plastic Bin
1/Pk.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Kappes Systeme™ Polystyrene Lid

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.