Learn More
Bochem™ Polystyrene Scapels
Brand:  Bochem™ 12640
Product Code. 15160216
  • 47.50 DKK / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Specifications
150 mm
Non-sterile
Polystyrene
Product Title
Bochem™ Polystyrene Scapels > Length: 150mm