Learn More
Polyurethane Stools
1405.00 DKK - 1455.00 DKK
Specifications
For Use With (Application) | For Office |
---|---|
Height (Metric) | 420 to 540 mm |
Quantity | 1 Ea. |
Product Type | Stool |
Material | Polyurethane |
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
11927664
|
mb2
AP4CB2D1NE2 |
5 Roulettes |
1455.00 DKK
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
11917664
|
mb2
AP4CB2D1NE3 |
5 Pads |
1405.00 DKK
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
Specifications
For Office | |
1 Ea. | |
Polyurethane |
420 to 540 mm | |
Stool |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Polyurethane Stools