missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Beige-Brown to White |
Physical Form | Crystalline Powder |
Linear Formula | (-CH2CH(OSO3K)-)n |
Quantity | 100 g |
IUPAC Name | potassium;ethenyl sulfate |
Percent Purity | ≥21.2% (K) |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Chemical Name or Material | Polyvinylsulfuric acid potassium salt |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000 >
Spot an opportunity for improvement?Share a Content Correction