Learn More
Fisherbrand™ Porcelain 25mL Crucibles
For laboratory and industrial purpose
124.00 DKK
Specifications
Color | White |
---|---|
Material | Porcelain |
For Use With (Application) | For laboratory and industrial purpose |
Capacity (Metric) | 25 mL |
Description
- Material type: Porcelain
- Color: White
- Cubic feet/case: 0.237
Specifications
White | |
Porcelain | |
For laboratory and industrial purpose | |
25 mL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Porcelain 25mL Crucibles