accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Mixers
Mortars and Pestles
Hecht Karl™ Porcelain Pestle
10489061
Print
Hecht Karl™ Porcelain Pestle
Outer Diameter: 130mm
Brand:
Hecht Karl™ 47031013
View more versions of this product
Product Code.
10489061
204.00 DKK
/ Each
Quantity
Estimated Shipment: 17-12-2024
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Specifications
Diameter (Metric) Outer
130mm
Material
Porcelain
Type
Pestle
Product Type
Pestle
Length (Metric)
150mm