missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Synonym: Chloro potassium
- Molecular Formula: KCl
- Molar Mass: 74.56g/mol
- CAS: 7447-40-7
- EIN ECS No.: 231-211-8
Specifications
Specifications
Boiling Point | 1413°C |
Melting Point | 773°C |
pH | 5.5–8.0 (5%, water) |
Quantity | 1 kg |
Formula Weight | 74.56g/mol |
Percent Purity | 99.5% min (argentometric) |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Chloride, Reagent Grade, Scharlab >
Spot an opportunity for improvement?Share a Content Correction