Learn More
Cole-Parmer™ Jenway™ Potassium Chloride Solution
Brand:  Cole-Parmer™ 68006-29
513.26 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11440529
  • 525.00 DKK / Each
Specifications
30mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ Jenway™ Potassium Chloride Solution > Capacity: 30mL