missing translation for 'onlineSavingsMsg'
Learn More
Potassium dihydrogen phosphate, Burdick & Jackson™ Product Code.: 15944888

Potassium dihydrogen phosphate, Burdick & Jackson™

Product Code. 15944888
1 kg, Plastic Bottle, 1kg
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 15944888

Brand: Honeywell-Burdick and Jackson 304071KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., Reagent ISO, Reagent European Pharmacopoeia

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point Decomposes on heating.
Melting Point 253°C
CAS 7778-77-0
Color Colorless
pH 4.2 to 4.5 (20°C)
Quantity 1 kg
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
Packaging Plastic Bottle
Chemical Name or Material Potassium dihydrogen phosphate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium dihydrogen phosphate, Burdick & Jackson™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.