missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide in Alcohol solution EU Pharmacopoeia 60%v/v, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Specifications
Specifications
Packaging | Bottle |
CAS | 1310-58-3, 7732-18-5 |
Molecular Formula | HKO |
Concentration | 0.5 M |
MDL Number | MFCD00003553 |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
SMILES | [OH-].[K+] |
IUPAC Name | potassium hydroxide |
Molecular Weight (g/mol) | 56.11 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
1121381 | Certificate of Analysis | 13/06/2025 | VL 039 1000 |
Safety and Handling


Product Identifier
- Potassium hydroxide 0.5M in alcohol (60% V/V). Volumetric Solution European Pharmacopoeia Ref: 3004900
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Hydroxide in Alcohol solution EU Pharmacopoeia 60%v/v, Fisher Chemical™ >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur