missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Potassium phosphate, 0.2M buffer soln., pH 8.0 Product Code.: 15433559

Thermo Scientific Chemicals Potassium phosphate, 0.2M buffer soln., pH 8.0

Product Code. 15433559
500 mL
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy

Product Code. 15433559

Brand: Thermo Scientific Chemicals J60851.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 500 mL
Chemical Name or Material Potassium phosphate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP19L552 Certificate TypeCertificate of Analysis Date07/05/2025 Catalog NumberJ60851.K2,J60851.AP
Lot NumberX23K529 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberJ60851.K2,J60851.AP
Lot NumberW21K513 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog NumberJ60851.K2,J60851.AP
Lot NumberR09K546 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog NumberJ60851.K2,J60851.AP
Lot NumberR09K545 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog NumberJ60851.K2,J60851.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Potassium phosphate, 0.2M buffer soln., pH 8.0 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.