missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Potassium phosphate monobasic, ACS Reagent, Honeywell Fluka™
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Potassium phosphate monobasic, ACS Reagent, Honeywell Fluka™
Product Code. 15657990 Shop All Honeywell Fluka Products2.5 kg
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
6 x 500 g
4 x 2.5 kg
This item is not returnable.
View return policy
Product Code. 15657990
Brand: Honeywell-Fluka P06622.5KG
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Chemical Identifiers
CAS | 7778-77-0 |
Molecular Formula | H2KO4P |
Molecular Weight (g/mol) | 136.08 |
MDL Number | MFCD00011401 MFCD00147253 |
InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphateShow MoreShow Less |
PubChem CID | 516951 |
ChEBI | CHEBI:63036 |
IUPAC Name | potassium;dihydrogen phosphate |
SMILES | [K+].OP(O)([O-])=O |
Specifications
Quantity | 2.5 kg |
Linear Formula | KH2PO4 |
UN Number | NONH for all modes of transport |
Formula Weight | 136.09g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Density | 2.338g/mL at 25°C |
Chemical Name or Material | Potassium phosphate monobasic |
Product Suggestions
You may be interested in these alternatives
-
Savings of 653.82 DKKProduct Code. 10409380
-
Savings of 541.32 DKKProduct Code. 10793611
-
Product Code. 10493465
-
Product Code. 15647990
-
Product Code. 15697990
-
Product Code. 15615940
-
Product Code. 10035810
-
Product Code. 15677990
-
Product Code. 15687990
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 3
-
Catalog No. 10703811
kr 1.480,00 / 500g -
Catalog No. 10793611
kr 525,00 / 1kg -
Catalog No. 10117113
kr 283,43 / 100g
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium phosphate monobasic, ACS Reagent, Honeywell Fluka™ >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction