Learn More
Memmert Potential-free contact (24V/2A) w/socket to NAMURNE 28, for signal ­ generation, for UN30pa
Brand:  Memmert UN30paH72
4275.13 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Potential-free contact (24V/2A) w/socket to NAMURNE 28, for signal ­ generation, for UN30pa
Product Code. 16324417
  • 4390.00 DKK / Each
Only null left
Estimated Shipment: 10-10-2025
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert Potential-free contact (24V/2A) w/socket to NAMURNE 28, for signal ­ generation, for UN30pa >