Learn More
PPP1CC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11082-1-AP-150UL
 View more versions of this product
Product Code. 16817923
  • 3058.99 DKK / 150µL
This item is not returnable. View return policy
Description
Description
DYRK1B is a member of the DYRK family of protein kinases. DYRK1B contains a bipartite nuclear localization signal and is found mainly in muscle and testis. The protein is proposed to be involved in the regulation of nuclear functions. Three isoforms of DYRK1B have been identified differing in the presence of two alternatively spliced exons within the catalytic domain.
Specifications
Specifications
PPP1CC
Polyclonal
Unconjugated
PPP1CC
PP1 Gamma, PP 1G, PPP1CC, PPP1G
Rabbit
Antigen Affinity Chromatography
RUO
19047, 24669, 5501
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P36873, P63087
Ppp1cc
PPP1CC Fusion Protein Ag1550
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PPP1CC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated