Learn More
Pre-Weigh Plate Count Agar (PCA)
Speed up your workflow when preparing medium for enumerating total viable counts in food, milk and dairy products with pre-weighed media sachets.
2378.89 DKK
Specifications
Description | Pre-Weigh Plate Count Agar (PCA) |
---|---|
Form | Powder |
Product Type | Agar |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15802477
|
Thermo Scientific Oxoid
PW0325-8L |
10 x Sachets for 8 L Medium | N/A | ||||||
Description
Speed up your workflow when preparing microbial culture medium for enumerating total viable counts (TVC) in food, milk and dairy products with Thermo Scientific™ Pre-weigh Plate Count Agar. The pre-weighed sachets of Thermo Scientific™ Oxoid™ dehydrated culture media are designed for single use when preparing 1 L or 8 L volumes of your medium. Save valuable time and start testing quickly by removing the requirement to weigh powder—just rip, tip and pour.
Pre-Weigh Plate Count Agar is equivalent to the medium recommended by the APHA1and the PHLS2 for the plate count of microorganisms in food, milk and other dairy products. It is part of a range of our most frequently used and well-characterized dehydrated culture media which is now available in handy pre-weighed foil packages.
Simply tear open the selected media sachet, add to the required volume of water, mix and autoclave as usual.
- Efficient: choose from a full range of pre-weighed media formats for reduced hands-on prep time and maximum productivity
- Convenient: rip and tip sachets enable faster media production and simplified workflows
- Compliant: proven and validated media formats deliver confidence and reliable performance
- Productive: reduce routine dehydrated culture media handling and weighing to spend more time on value-added tasks.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Pre-Weigh Plate Count Agar (PCA) | |
Powder | |
Agar |
For Laboratory Use.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.