Learn More
Kern Sohn Precision Balance PLS
High-quality precision scale with comfortable graphic display and enormous weighing range.
Brand:  Kern Sohn PLS 4200-2F
Encompass_Preferred
 View more versions of this product
Product Code. 17351743
This item is not returnable. View return policy
Description
Description
  • Convenient recipe-weighing: with the recipe database, in which up to 99 recipes can be stored, each with up to 20 recipe ingredients with name and target value
  • Rapid and efficient operation thanks to the graphics display
  • Simple, clear user interface on the display in the following languages: German, English, French, Italian, Spanish, Portuguese
  • Dosage aid: High stability mode and other filter settings can be selected
  • Internal memory for complete recipes with name and target value of the recipe ingredients. User guidance through display
  • Protective working cover included with delivery
Specifications
Specifications
4200 g
0.04 g
Universal Mains Adapter
0.01 g
Precision Balance