Learn More
Kern Sohn Precision Balance PLS
High-quality precision scale with comfortable graphic display and enormous weighing range.
Brand:  Kern Sohn PLS 720-3A
Encompass_Preferred
 View more versions of this product
Product Code. 17371743
  • 8903.09 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Convenient recipe-weighing: with the recipe database, in which up to 99 recipes can be stored, each with up to 20 recipe ingredients with name and target value
  • Rapid and efficient operation thanks to the graphics display
  • Simple, clear user interface on the display in the following languages: German, English, French, Italian, Spanish, Portuguese
  • Dosage aid: High stability mode and other filter settings can be selected
  • Internal memory for complete recipes with name and target value of the recipe ingredients. User guidance through display
  • Protective working cover included with delivery
Specifications
Specifications
720 g
0.002 g
Universal Mains Adapter
0.001 g
Precision Balance
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.