Learn More
Kern Sohn Precision Balance PLS
High-quality precision scale with comfortable graphic display and enormous weighing range.
Brand: Kern Sohn PLS 8000-2A
This item is not returnable.
View return policy
Description
- Convenient recipe-weighing: with the recipe database, in which up to 99 recipes can be stored, each with up to 20 recipe ingredients with name and target value
- Rapid and efficient operation thanks to the graphics display
- Simple, clear user interface on the display in the following languages: German, English, French, Italian, Spanish, Portuguese
- Dosage aid: High stability mode and other filter settings can be selected
- Internal memory for complete recipes with name and target value of the recipe ingredients. User guidance through display
- Protective working cover included with delivery
Specifications
8200 g | |
0.04 g | |
Universal Mains Adapter |
0.01 g | |
Precision Balance |