Learn More
Bochem™ Precision Forceps, PTFE
Sharply pointed precision forceps. Bochem™ Precision Forceps, PTFE are coated with PTFE for greater resistance.
Brand: Bochem™ 1860
This item is not returnable.
View return policy
Description
- Precision style.
- Extra sharp point design.
- Points without ridges.
- PTFE-coated.
Specifications
Extra sharp, straight, unridged | |
PTFE-coated | |
Forceps |
Gradual taper | |
120 mm |