Learn More
Bochem™ Precision Forceps, PTFE
Sharply pointed precision forceps. Bochem™ Precision Forceps, PTFE are coated with PTFE for greater resistance.
Brand:  Bochem™ 1865
 View more versions of this product
Product Code. 10480431
  • 232.00 DKK / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
Description
Description
  • Precision style.
  • Extra sharp point design.
  • Points without ridges.
  • PTFE-coated.
Specifications
Specifications
Extra sharp, straight, unridged
PTFE-coated
Forceps
Broad handle, fast taper
105 mm