Learn More
Invitrogen™ PRIMER, 5'ALEXA 405
Custom Primer
Brand:  Invitrogen™ 100007329
1065.00 DKK valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10232759
  • 1420.00 DKK / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' Alexa 405 Primer, 50 nmol
5' Alexa 405 Primer
5' Labeled
50 nmol