Learn More
Invitrogen™ PRIMER 5'PHO 25NMOLE PLATE
Custom Primer
Brand:  Invitrogen™ 10625101
47.06 DKK valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10378072
  • 62.75 DKK / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' PHO Primer Plate, 25 nmol
5' PHO Primer Plate
5' Labeled
25 nmol