Learn More
Invitrogen™ PRIMER URACIL 1 UMOL
Custom Primer
Brand:  Invitrogen™ 10623023
231.75 DKK valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10164042
  • 309.00 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Uracil Primer, 1μmol
Uracil Primer
1 μmol