Learn More
Thermo Scientific™ Printers for CryoMed™ and CryoPlus™ Freezers
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with this optional printer.
Brand: Thermo Scientific™ 4000566
1517.27 DKK valid until 2025-08-22
Use promo code "24148" to get your promotional price.
Description
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with Thermo Scientific™ Thermal Printers for CryoMed and CryoPlus™ Freezers. These printers are available in 120V or 220V models, and include cable assembly and RS-232 connector kit.
- Model 4000565 - 120V, 60Hz printer with cable assembly and RS-232 connector kit
- Model 4000665 - 220V, 50Hz printer with cable assembly and RS-232 connector kit
- Model 4000566 - Thermal printer paper (5 rolls/package)
Specifications
Thermal Printer Paper | |
All CryoMed Controlled-Rate Freezers | |
Not Applicable |
Promotions
-
Multiple Manufacturers
Illuminating Protein Pathways
Shining a light on the tools that drive protein discovery.Promo Code: 24148Expires: 2025-08-22
New Offer Save Now