Learn More
Bollé Safety Prism Clear Safety Glasses
Manufactured by Bollé Safety, Prism Safety Glasses offer excellent protection and are great value. They feature a durable polycarbonate frame, with an anti-scratch lens coating.
Brand: Bollé Safety PRIPSI
44.39 DKK valid until 2025-06-08
Use promo code "24131" to get your promotional price.
This item is not returnable.
View return policy
Description
- Upper protection
- Sporty design
- Comfortable, straight temples
- Anti-fog & anti-scratch coating
Specifications
Anti-fog, Scratch-resistant | |
Prism Safety Glasses | |
1 Ea. |
Clear | |
Adjustable cord | |
Polycarbonate |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Bollé Safety Prism Clear Safety Glasses > Frame Material: Polycarbonate
Spot an opportunity for improvement?Share a Content Correction