missing translation for 'onlineSavingsMsg'
Learn More

PRMT2 Mouse anti-Human, Unconjugated, Clone: 1F1B9, Proteintech

Mouse Monoclonal Antibody

1081.84 DKK - 3058.99 DKK

Specifications

Antigen PRMT2
Clone 1F1B9
Concentration 1000 μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16869155
View Documents
Proteintech
66885-1-IG-20UL
20 μL
1081.84 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
16859155
View Documents
Proteintech
66885-1-IG-150UL
150 μL
3058.99 DKK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The PRMT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRMT2. This antibody recognizes human, mouse, rat antigen. The PRMT2 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

PRMT2
1000 μg/mL
Monoclonal
Mouse
3275
Primary
1F1B9
Western Blot
Unconjugated
Human, Rat, Mouse
IgG2a
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PRMT2 Mouse anti-Human, Unconjugated, Clone: 1F1B9, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.