Learn More
Invitrogen™ ProcartaPlex™ Flat Bottom Plate, 96-well
This black ProcartaPlex 96-well flat-bottom plate is ideal for use with all ProcartaPlex Luminex protein assays.
Brand:  Invitrogen™ EPX-88182-000
Product Code. 18739279
  • 153.00 DKK / Each
Explore more special offers
Description
Description
This black ProcartaPlex 96-well flat-bottom plate is ideal for use with all ProcartaPlex Luminex protein assays.
This plate is included in all ProcartaPlex multiplex panel kits, species-specific basic kits, and as part of custom mix and match panels. For bulk orders, please contact your sales representative.
Note that the reusable lid (Cat. No. EPX-88104-000) is sold separately.
About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to detect and quantitate up to 80 protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.
ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, non-human primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.
Related products
  • ProcartaPlex Microplate Lid (Cat. No. EPX-88104-000)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96-well
ProcartaPlex
Single-use
Polystyrene
Non-sterile
Flat Bottom (F-well)
Certified Quality
Without Lid
Black
Plate
96
Hydrophobic
1 Plate