Learn More
Invitrogen™ ProcartaPlex™ Microplate Lid
This reusable microplate lid is ideal for use with all ProcartaPlex Luminex protein assays.
Brand:  Invitrogen™ EPX-88104-000
201.40 DKK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 17896527
  • 219.00 DKK / Pack of 96
Explore more special offers
Description
Description
This reusable microplate lid is ideal for use with all ProcartaPlex Luminex protein assays.
This lid is included in all ProcartaPlex multiplex panel kits, species-specific basic kits, and as part of custom mix and match panels. For bulk orders, please contact your sales representative.
Note that the 96-well flat bottom plate (Cat. No. EPX-88182-000) is sold separately.
About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to detect and quantitate up to 80 protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.
ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, non-human primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.
Related products
  • ProcartaPlex Flat Bottom Plate, 96-well (Cat. No. EPX-88182-000)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
Non-sterile
96 tests
Certified Quality
Polystyrene
96
Plate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number445524-000 Certificate TypeCertificate of Analysis Date28/03/2025 Catalog Number
Lot Number443938-000 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog Number
Lot Number442470-000 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog Number
Lot Number439488-000 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog Number
Lot Number433305-000 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog NumberEPX-88104-000
Product Title
Invitrogen™ ProcartaPlex™ Microplate Lid > 1 Lid