Learn More
Invitrogen™ eBioscience™ ProcartaPlex Rat MIP-2 alpha (CXCL2) Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand: Invitrogen™ EPX01A-36054-901
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to rat MIP-2
- rat MIP-2 Standard (lyophilized)
- Biotin-Conjugate (50x) anti-rat MIP-2 monoclonal antibody
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
96 Tests | |
Multiplex Immunoassay | |
Cell Culture Supernatant, Plasma, Serum | |
Gro beta |
Rat | |
CXCL2 (MIP-2) | |
Serum, Plasma: 25 μL; CCS: 50 μL |