Learn More
uvex ProfiErgo ENB 20 Nitrile-Coated Assembly Gloves
Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
Description
- Thick, special NBR coating is resistant to oil and grease
- High-quality cotton interlock liner is comfortable, feels pleasant against the skin, and aids in regulation of the glove climate
- Enhanced grip capabilities in damp, wet, and oily areas
- Gloves are suitable for operating touch displays
- Ideal for a variety of applications including assembly, control work, and maintenance
Specifications
Specifications
Coating Material | NBR |
Color | White, Yellow |
Exterior Finish | Smooth |
Hand | Ambidextrous |
Latex Free | Yes |
Material | Cotton |
Product Type | Mechanical Protection Gloves |
Size | 6 |
EN388 Standard | 2121X |
Glove Cuff Style | Knit |
Show More |
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
uvex ProfiErgo ENB 20 Nitrile-Coated Assembly Gloves >
Spot an opportunity for improvement?Share a Content Correction