Learn More
Bochem™ Propeller Stirrers
Mixes media with low and middle viscosity
Brand:  Bochem™ 9114
Product Code. 10449582
  • 279.00 DKK / Each
Only null left
Estimated Shipment: 19-08-2025
Description
  • Three fixed blades
  • Rpm-range middle
  • Mixing of media with low and middle viscosity
  • Ideal for homogenising and suspensioning
  • Axial flow
Specifications
Specifications
Stainless Steel (18/8)
Ideal for Homogenizing and Suspensieren
1/Pk.
Propeller Stirrers
500 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Propeller Stirrers > Blade Length: 500mm